siRNA Details
| Gene Symbol: Cntn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159647.1 | NP_001153119.1 | 5 | 639 | ||
| NM_001159648.1 | NP_001153120.1 | 5 | 493 | |||
| NM_007727.2 | NP_031753.1 | 5 | 542 | |||
| GenBank mRNA | AK138969.1 | - | 5 | 494 | ||
| BC066864.1 | - | 5 | 565 | 30099512 | ||
| X14943.1 | - | 5 | 512 | |||

Cntn1
Genomic Map