siRNA Details
| Gene Symbol: Cox6a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007748.3 | NP_031774.1 | 3 | 333 | ||
| GenBank mRNA | AK002402.1 | - | 3 | 335 | ||
| AK019318.1 | - | 3 | 338 | |||
| AK166496.1 | - | 3 | 335 | |||
| BC003807.1 | - | 3 | 333 | 3487598 | ||
| BC052816.1 | - | 3 | 339 | 6439987 | ||
| L06465.1 | - | 3 | 332 | |||
| U08440.1 | - | 3 | 322 | |||

Cox6a1
Genomic Map