siRNA Details
| Gene Symbol: Cpeb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252525.1 | NP_001239454.1 | 4 | 605 | ||
| NM_001252526.1 | NP_001239455.1 | 4 | 602 | |||
| NM_007755.5 | NP_031781.1 | 4 | 602 | |||
| GenBank mRNA | AK136088.1 | - | 4 | 605 | ||
| BC125476.1 | - | 4 | 522 | 40129991 | ||
| BC144948.1 | - | 4 | 519 | |||
| BY720025.1 | - | 4 | 605 | |||
| Y08260.1 | - | 4 | 521 | |||

Cpeb1
Genomic Map