siRNA Details
| Gene Symbol: Cr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007758.2 | NP_031784.1 | 4 | 815 | ||
| GenBank mRNA | AK003072.2 | - | 4 | 815 | ||
| BC109344.1 | - | 4 | 748 | 40054528 | ||
| M35684.1 | - | 3 | 683 | |||
| M61132.1 | - | 3 | 650 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cr2| Gene Symbol: Cr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007758.2 | NP_031784.1 | 4 | 815 | ||
| GenBank mRNA | AK003072.2 | - | 4 | 815 | ||
| BC109344.1 | - | 4 | 748 | 40054528 | ||
| M35684.1 | - | 3 | 683 | |||
| M61132.1 | - | 3 | 650 | |||