siRNA Details
| Gene Symbol: Crx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113330.1 | NP_001106801.1 | 4 | 1595 | ||
| NM_007770.4 | NP_031796.1 | 4 | 1623 | |||
| GenBank mRNA | AK053533.1 | - | 4 | 1565 | ||
| AK139691.1 | - | 4 | 1598 | |||
| BC016502.1 | - | 4 | 1537 | 4527863 | ||

Crx
Genomic Map