siRNA Details
| Gene Symbol: Ctse | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007799.3 | NP_031825.2 | 6 | 892 | ||
| GenBank mRNA | AK143581.1 | - | 5 | 594 | ||
| AK145875.1 | - | 6 | 749 | |||
| AK157907.1 | - | 5 | 614 | |||
| AK165271.1 | - | 5 | 560 | |||
| BC005432.1 | - | 6 | 892 | 3601519 | ||
| CT010252.1 | - | 5 | 528 | |||
| X97399.1 | - | 5 | 579 | |||

Ctse
Genomic Map