siRNA Details
| Gene Symbol: Cybb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007807.5 | NP_031833.3 | 5 | 487 | ||
| GenBank mRNA | AK033570.1 | - | 5 | 431 | ||
| AK053920.1 | - | 5 | 431 | |||
| AK152257.1 | - | 5 | 458 | |||
| AK153154.1 | - | 5 | 458 | |||
| AK153259.1 | - | 5 | 458 | |||
| AK154126.1 | - | 5 | 487 | |||
| AK171526.1 | - | 5 | 473 | |||
| BC042838.1 | - | 5 | 454 | |||
| BC071229.1 | - | 5 | 442 | 4979804 | ||
| FJ168469.1 | - | 5 | 476 | |||
| U43384.1 | - | 5 | 413 | |||

Cybb
Genomic Map