siRNA Details
| Gene Symbol: Cybb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007807.5 | NP_031833.3 | 13 | 1703 | ||
| GenBank mRNA | AK033570.1 | - | 13 | 1648 | ||
| AK053920.1 | - | 13 | 1647 | |||
| AK152257.1 | - | 13 | 1674 | |||
| AK153071.1 | - | 6 | 813 | |||
| AK153154.1 | - | 13 | 1674 | |||
| AK153259.1 | - | 13 | 1674 | |||
| AK154126.1 | - | 13 | 1703 | |||
| AK160031.1 | - | 8 | 1105 | |||
| AK171526.1 | - | 13 | 1689 | |||
| BC003910.1 | - | 4 | 392 | |||
| BC071229.1 | - | 13 | 1658 | 4979804 | ||
| FJ168469.1 | - | 13 | 1692 | |||
| U43384.1 | - | 13 | 1629 | |||

Cybb
Genomic Map