siRNA Details
| Gene Symbol: Cyp2b19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF047529.1 | - | 9 | 1418 | ||
| BC137965.1 | - | 9 | 1463 | 8860984 | ||
| BC137966.1 | - | 9 | 1463 | 8860985 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cyp2b19| Gene Symbol: Cyp2b19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF047529.1 | - | 9 | 1418 | ||
| BC137965.1 | - | 9 | 1463 | 8860984 | ||
| BC137966.1 | - | 9 | 1463 | 8860985 | ||