siRNA Details
| Gene Symbol: Cyp4a14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007822.2 | NP_031848.1 | 9 | 1169 | ||
| GenBank mRNA | AK098088.1 | - | 9 | 1166 | ||
| AK149392.1 | - | 9 | 1167 | |||
| AK165452.1 | - | 9 | 1167 | |||
| BC089609.1 | - | 9 | 1169 | |||

Cyp4a14
Genomic Map