siRNA Details
| Gene Symbol: Dcc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007831.3 | NP_031857.2 | 20 | 3678 | ||
| GenBank mRNA | AK157715.1 | - | 20 | 3567 | ||
| X85788.1 | - | 17 | 3228 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dcc| Gene Symbol: Dcc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007831.3 | NP_031857.2 | 20 | 3678 | ||
| GenBank mRNA | AK157715.1 | - | 20 | 3567 | ||
| X85788.1 | - | 17 | 3228 | |||