siRNA Details
| Gene Symbol: PI4KA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_058004.3 | NP_477352.3 | 37, 38 | 4503 | ||
| XM_005261634.1 | XP_005261691.1 | 36, 37 | 4410 | |||
| XM_017028829.1 | XP_016884318.1 | 36, 37 | 4437 | |||
| XM_017028830.1 | XP_016884319.1 | 23, 24 | 2659 | |||
| GenBank mRNA | AB210002.1 | - | 37, 38 | 4473 | ||
| AF012872.1 | - | 36, 37 | 4242 | |||
| AK302424.1 | - | 37, 38 | 4431 | |||
| AK314781.1 | - | 9, 10 | 1157 | |||
| BC018120.1 | - | 7, 8 | 820 | |||
| BC053654.1 | - | 6, 7 | 796 | |||
| L36151.1 | - | 5, 6 | 789 | |||

PI4KA
Genomic Map