siRNA Details
| Gene Symbol: Dhx9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007842.2 | NP_031868.2 | 3, 4 | 402 | ||
| GenBank mRNA | AK013423.1 | - | 3, 4 | 403 | ||
| AK141824.1 | - | 3, 4 | 394 | |||
| AK169735.1 | - | 3, 4 | 402 | |||
| BC033911.1 | - | 3, 4 | 402 | |||
| BC089159.1 | - | 3, 4 | 396 | |||
| U91922.1 | - | 3, 4 | 359 | |||

Dhx9
Genomic Map