siRNA Details
| Gene Symbol: Twist2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007855.2 | NP_031881.1 | 2 | 1292 | ||
| GenBank mRNA | AK008411.1 | - | 2 | 1029 | ||
| AK011180.1 | - | 2 | 1308 | |||
| AK011861.1 | - | 2 | 1309 | |||
| BC090636.1 | - | 2 | 1297 | 30942245 | ||
| U36384.1 | - | 2 | 1293 | |||

Twist2
Genomic Map