siRNA Details
Gene Symbol: Dhh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007857.4 | NP_031883.1 | 3 | 2003 | ||
GenBank mRNA | AK032466.1 | - | 3 | 1987 | ||
AK033392.1 | - | 3 | 2005 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Dhh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007857.4 | NP_031883.1 | 3 | 2003 | ||
GenBank mRNA | AK032466.1 | - | 3 | 1987 | ||
AK033392.1 | - | 3 | 2005 |