siRNA Details
| Gene Symbol: Diap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK140734.1 | - | 5 | 484 | ||
| AK220466.1 | - | 5 | 493 | |||
| BC021396.1 | - | 4 | 523 | |||
| BC070412.1 | - | 6 | 658 | 5705112 | ||
| U96963.1 | - | 4 | 496 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Diap1| Gene Symbol: Diap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK140734.1 | - | 5 | 484 | ||
| AK220466.1 | - | 5 | 493 | |||
| BC021396.1 | - | 4 | 523 | |||
| BC070412.1 | - | 6 | 658 | 5705112 | ||
| U96963.1 | - | 4 | 496 | |||