siRNA Details
| Gene Symbol: NEU2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005383.2 | NP_005374.2 | 2 | 1088 | ||
| GenBank mRNA | BC069151.1 | - | 2 | 1088 | 7217011 | |
| BC107053.2 | - | 2 | 1088 | 40004620 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

NEU2| Gene Symbol: NEU2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005383.2 | NP_005374.2 | 2 | 1088 | ||
| GenBank mRNA | BC069151.1 | - | 2 | 1088 | 7217011 | |
| BC107053.2 | - | 2 | 1088 | 40004620 | ||