siRNA Details
| Gene Symbol: Dll1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007865.3 | NP_031891.2 | 11 | 3061 | ||
| GenBank mRNA | BC057400.1 | - | 11 | 2574 | 6402691 | |
| BC065063.1 | - | 11 | 2825 | 6416191 | ||
| X80903.1 | - | 11 | 2491 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dll1| Gene Symbol: Dll1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007865.3 | NP_031891.2 | 11 | 3061 | ||
| GenBank mRNA | BC057400.1 | - | 11 | 2574 | 6402691 | |
| BC065063.1 | - | 11 | 2825 | 6416191 | ||
| X80903.1 | - | 11 | 2491 | |||