siRNA Details
| Gene Symbol: Dll3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007866.2 | NP_031892.2 | 7 | 1679 | ||
| GenBank mRNA | AB013440.1 | - | 7 | 1666 | ||
| AK144282.1 | - | 7 | 1680 | |||
| BC052002.1 | - | 7 | 1656 | 6404029 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dll3| Gene Symbol: Dll3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007866.2 | NP_031892.2 | 7 | 1679 | ||
| GenBank mRNA | AB013440.1 | - | 7 | 1666 | ||
| AK144282.1 | - | 7 | 1680 | |||
| BC052002.1 | - | 7 | 1656 | 6404029 | ||