siRNA Details
| Gene Symbol: Dnajc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001190817.1 | NP_001177746.1 | 1 | 482 | ||
| NM_007869.3 | NP_031895.1 | 1 | 482 | |||
| GenBank mRNA | AK012505.1 | - | 1 | 476 | ||
| AK052022.1 | - | 1 | 152 | |||
| AK133582.1 | - | 1 | 466 | |||
| AK144228.1 | - | 1 | 438 | |||
| BC020071.1 | - | 367 | ||||
| BC080300.1 | - | 1 | 463 | |||
| L16953.1 | - | 1 | 159 | |||

Dnajc1
Genomic Map