siRNA Details
| Gene Symbol: Reep5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007874.3 | NP_031900.3 | 2 | 282 | ||
| GenBank mRNA | AK010507.1 | - | 2 | 234 | ||
| AK013146.1 | - | 2 | 213 | |||
| AK151058.1 | - | 2 | 237 | |||
| AK151271.1 | - | 2 | 237 | |||
| AK152361.1 | - | 2 | 211 | |||
| AK152503.1 | - | 2 | 237 | |||
| AK167385.1 | - | 2 | 211 | |||
| AY562233.1 | - | 2 | 188 | |||
| BC013052.1 | - | 2 | 213 | 3585290 | ||
| U28168.1 | - | 2 | 211 | |||

Reep5
Genomic Map