siRNA Details
| Gene Symbol: Reep5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007874.3 | NP_031900.3 | 5 | 1034 | ||
| GenBank mRNA | AK013146.1 | - | 5 | 965 | ||
| BC013052.1 | - | 5 | 965 | 3585290 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Reep5| Gene Symbol: Reep5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007874.3 | NP_031900.3 | 5 | 1034 | ||
| GenBank mRNA | AK013146.1 | - | 5 | 965 | ||
| BC013052.1 | - | 5 | 965 | 3585290 | ||