siRNA Details
Gene Symbol: Drg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007879.1 | NP_031905.1 | 3 | 271 | ||
GenBank mRNA | AK044194.1 | - | 3 | 280 | ||
AK080814.1 | - | 3 | 269 | |||
AK090156.1 | - | 3 | 301 | |||
AK144438.1 | - | 3 | 273 | |||
AK155061.1 | - | 3 | 286 | |||
AK208688.1 | - | 2 | 147 | |||
AK210426.1 | - | 2 | 46 | |||
AK211821.1 | - | 2 | 46 | |||
BC039649.1 | - | 3 | 265 | 5351885 | ||
D10715.1 | - | 3 | 271 |
