siRNA Details
| Gene Symbol: Slc26a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007885.2 | NP_031911.1 | 4 | 1612 | ||
| GenBank mRNA | AK052942.1 | - | 4 | 1613 | ||
| AK163974.1 | - | 4 | 1503 | |||
| AK165805.1 | - | 2 | 919 | |||
| BC028345.1 | - | 4 | 1489 | 4014785 | ||
| D42049.1 | - | 4 | 1497 | |||

Slc26a2
Genomic Map