siRNA Details
| Gene Symbol: Dtnb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001162465.1 | NP_001155937.1 | 10, 11 | 1345 | ||
| NM_007886.2 | NP_031912.2 | 10, 11 | 1280 | |||
| GenBank mRNA | AJ003007.1 | - | 10, 11 | 1213 | ||
| AK043436.1 | - | 10, 11 | 1303 | |||
| AK188811.1 | - | 3, 4 | 329 | |||
| BC030924.1 | - | 10, 11 | 1280 | 4910166 | ||
| Y15742.1 | - | 10, 11 | 1177 | |||

Dtnb
Genomic Map