siRNA Details
| Gene Symbol: CXCL11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001302123.1 | NP_001289052.1 | 3 | 429 | ||
| NM_005409.4 | NP_005400.1 | 3 | 429 | |||
| GenBank mRNA | AF002985.1 | - | 3 | 306 | ||
| AF030514.1 | - | 3 | 288 | |||
| AL703031.1 | - | 3 | 269 | |||
| BC005292.1 | - | 3 | 279 | 3934139 | ||
| BC110986.1 | - | 3 | 288 | 5178174 | ||
| BP289021.1 | - | 3 | 331 | |||
| BT006787.1 | - | 3 | 219 | |||
| U66096.1 | - | 3 | 264 | |||
| Y15220.1 | - | 3 | 225 | |||

CXCL11
Genomic Map