siRNA Details
| Gene Symbol: Ankfy1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009671.5 | NP_033801.4 | 5 | 556 | ||
| GenBank mRNA | AB011370.2 | - | 5 | 554 | ||
| AB098329.1 | - | 5 | 554 | |||
| BC139231.1 | - | 5 | 567 | 8862253 | ||
| BC145583.1 | - | 5 | 567 | 9054149 | ||
| BQ550282.1 | - | 5 | 561 | |||

Ankfy1
Genomic Map