siRNA Details
| Gene Symbol: Ankfy1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009671.5 | NP_033801.4 | 25 | 5304 | ||
| GenBank mRNA | AB011370.2 | - | 25 | 5346 | ||
| AB098329.1 | - | 25 | 5297 | |||
| AK014592.1 | - | 1 | 1664 | |||
| AK122479.1 | - | 5 | 2303 | |||
| BC139231.1 | - | 25 | 5315 | 8862253 | ||
| BC145583.1 | - | 25 | 5310 | 9054149 | ||

Ankfy1
Genomic Map