siRNA Details
| Gene Symbol: Ap1g1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001301211.1 | NP_001288140.1 | 16, 17 | 2017 | ||
| NM_009677.6 | NP_033807.2 | 17, 18 | 2026 | |||
| GenBank mRNA | AK028360.1 | - | 5, 6 | 432 | ||
| AK034198.1 | - | 14, 15 | 1255 | |||
| AK036387.1 | - | 17, 18 | 1969 | |||
| AK145821.1 | - | 16, 17 | 1948 | |||
| BC006691.1 | - | 16, 17 | 1693 | |||
| BC052703.1 | - | 17, 18 | 1899 | 5698075 | ||
| BC054535.1 | - | 17, 18 | 1899 | 5698075 | ||
| X54424.1 | - | 16, 17 | 1939 | |||

Ap1g1
Genomic Map