siRNA Details
| Gene Symbol: TECTA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005422.2 | NP_005413.2 | 2 | 82 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_009347, BC171953, BC138426, X99805.

TECTA
Genomic Map