siRNA Details
| Gene Symbol: Araf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009703.2 | NP_033833.1 | 16 | 2176 | ||
| GenBank mRNA | AK034861.1 | - | 17 | 2283 | ||
| AK085823.1 | - | 16 | 2144 | |||
| BC004757.1 | - | 16 | 2132 | 3154293 | ||
| D00024.1 | - | 11 | 1538 | |||

Araf
Genomic Map