siRNA Details
| Gene Symbol: TFF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005423.4 | NP_005414.1 | 3 | 402 | ||
| GenBank mRNA | BC032820.1 | - | 3 | 281 | 5184770 | |
| BM817851.1 | - | 3 | 301 | |||
| BM853946.1 | - | 3 | 299 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TFF2| Gene Symbol: TFF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005423.4 | NP_005414.1 | 3 | 402 | ||
| GenBank mRNA | BC032820.1 | - | 3 | 281 | 5184770 | |
| BM817851.1 | - | 3 | 301 | |||
| BM853946.1 | - | 3 | 299 | |||