siRNA Details
| Gene Symbol: Bag1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001171739.1 | NP_001165210.1 | 2 | 591 | ||
| NM_009736.3 | NP_033866.4 | 2 | 591 | |||
| GenBank mRNA | AF022223.1 | - | 2 | 532 | ||
| AK009149.1 | - | 2 | 502 | |||
| AK154469.1 | - | 2 | 541 | |||
| AK171094.1 | - | 2 | 541 | |||
| BC003722.1 | - | 2 | 553 | 3495306 | ||
| BC069918.1 | - | 2 | 528 | 4979705 | ||
| BC093509.1 | - | 2 | 502 | 4978330 | ||
| CA321856.1 | - | 2 | 578 | |||
| CX225744.1 | - | 2 | 591 | |||

Bag1
Genomic Map