siRNA Details
| Gene Symbol: Bmx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009759.4 | NP_033889.2 | 4 | 456 | ||
| GenBank mRNA | AF012104.1 | - | 3 | 309 | ||
| AK036707.1 | - | 3 | 315 | |||
| AK040936.1 | - | 4 | 457 | |||
| AK080038.1 | - | 4 | 457 | |||
| BC138104.1 | - | 4 | 333 | 8861123 | ||
| BC138105.1 | - | 4 | 333 | 8861124 | ||
| U88091.1 | - | 4 | 442 | |||

Bmx
Genomic Map