siRNA Details
| Gene Symbol: Bmx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009759.4 | NP_033889.2 | 19 | 2341 | ||
| GenBank mRNA | AF012104.1 | - | 18 | 2194 | ||
| AK036707.1 | - | 18 | 2200 | |||
| AK040936.1 | - | 19 | 2342 | |||
| BC138104.1 | - | 19 | 2218 | 8861123 | ||
| BC138105.1 | - | 19 | 2218 | 8861124 | ||
| U88091.1 | - | 19 | 2327 | |||

Bmx
Genomic Map