siRNA Details
Gene Symbol: Smyd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160127.1 | NP_001153599.1 | 1, 2 | 222 | ||
NM_009762.2 | NP_033892.2 | 1, 2 | 222 | |||
GenBank mRNA | AK076266.1 | - | 1, 2 | 186 | ||
AK142153.1 | - | 1, 2 | 190 | |||
AK142252.1 | - | 1, 2 | 200 | |||
BC076601.1 | - | 1, 2 | 200 | 30609878 | ||
BY089666.1 | - | 1, 2 | 223 | |||
U76373.2 | - | 1, 2 | 194 | |||
U76374.2 | - | 1, 2 | 194 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
