siRNA Details
| Gene Symbol: Brca1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009764.3 | NP_033894.3 | 23 | 5711 | ||
| GenBank mRNA | BC068303.1 | - | 23 | 5544 | 30431022 | |
| U31625.1 | - | 23 | 5568 | |||
| U36475.1 | - | 22 | 5531 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Brca1| Gene Symbol: Brca1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009764.3 | NP_033894.3 | 23 | 5711 | ||
| GenBank mRNA | BC068303.1 | - | 23 | 5544 | 30431022 | |
| U31625.1 | - | 23 | 5568 | |||
| U36475.1 | - | 22 | 5531 | |||