siRNA Details
| Gene Symbol: Bsg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009768.2 | NP_033898.1 | 2 | 346 | ||
| GenBank mRNA | AK147225.1 | - | 2 | 4046 | ||
| AY089967.1 | - | 2 | 204 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bsg| Gene Symbol: Bsg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009768.2 | NP_033898.1 | 2 | 346 | ||
| GenBank mRNA | AK147225.1 | - | 2 | 4046 | ||
| AY089967.1 | - | 2 | 204 | |||