siRNA Details
| Gene Symbol: C3ar1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009779.2 | NP_033909.1 | 2 | 1132 | ||
| GenBank mRNA | AK054017.1 | - | 2 | 1135 | ||
| AK154229.1 | - | 2 | 1120 | |||
| AK154609.1 | - | 2 | 1059 | |||
| AY786554.1 | - | 1 | 950 | |||
| BC003728.1 | - | 2 | 1045 | 3593758 | ||
| U77460.1 | - | 2 | 980 | |||
More Information
Set Membership: |
|

C3ar1
Genomic Map