siRNA Details
| Gene Symbol: ERCC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005236.2 | NP_005227.1 | 1, 2 | 224 | ||
| XM_011522424.2 | XP_011520726.1 | 1, 2 | 243 | |||
| XM_017023043.1 | XP_016878532.1 | 1, 2 | 190 | |||
| GenBank mRNA | AK289726.1 | - | 1, 2 | 223 | ||
| AK308341.1 | - | 1, 2 | 190 | |||
| BC020741.1 | - | 1, 2 | 195 | |||
| BC142631.1 | - | 1, 2 | 224 | |||
| U64315.1 | - | 1, 2 | 197 | |||

ERCC4
Genomic Map