siRNA Details
| Gene Symbol: Casp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001284409.1 | NP_001271338.1 | 7 | 1521 | ||
| NM_009810.3 | NP_033940.1 | 7 | 1516 | |||
| GenBank mRNA | AK049043.1 | - | 1 | 437 | ||
| AK080651.1 | - | 4 | 1031 | |||
| AK146226.1 | - | 7 | 1522 | |||
| BC038825.1 | - | 7 | 1405 | 4482368 | ||
| U19522.1 | - | 7 | 1332 | |||
| U49929.1 | - | 7 | 1358 | |||
| U63720.1 | - | 4 | 1123 | |||

Casp3
Genomic Map