siRNA Details
| Gene Symbol: GEM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005261.3 | NP_005252.1 | 2, 3 | 568 | ||
| NM_181702.2 | NP_859053.1 | 2, 3 | 453 | |||
| XM_017013315.1 | XP_016868804.1 | 2, 3 | 594 | |||
| GenBank mRNA | AK314017.1 | - | 2, 3 | 453 | ||
| BC022010.2 | - | 2, 3 | 453 | 4823985 | ||
| BI759640.1 | - | 2, 3 | 463 | |||
| U10550.1 | - | 2, 3 | 542 | |||
| U13052.1 | - | 1, 2 | 329 | |||

GEM
Genomic Map