siRNA Details
| Gene Symbol: Entpd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF037366.1 | - | 5 | 496 | ||
| AK028625.1 | - | 2 | 162 | |||
| AK029512.1 | - | 5 | 542 | |||
| AK030601.1 | - | 5 | 522 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Entpd1| Gene Symbol: Entpd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF037366.1 | - | 5 | 496 | ||
| AK028625.1 | - | 2 | 162 | |||
| AK029512.1 | - | 5 | 542 | |||
| AK030601.1 | - | 5 | 522 | |||