siRNA Details
| Gene Symbol: Cd7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009854.2 | NP_033984.1 | 2 | 368 | ||
| GenBank mRNA | AK136444.1 | - | 2 | 322 | ||
| AK154339.1 | - | 2 | 323 | |||
| BC024376.1 | - | 2 | 309 | 4985794 | ||
| CT010368.1 | - | 2 | 255 | |||
| D10329.1 | - | 2 | 308 | |||

Cd7
Genomic Map