siRNA Details
| Gene Symbol: Cdc25c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK141737.1 | - | 9 | 958 | ||
| AK181318.1 | - | 6 | 352 | |||
| BC004669.1 | - | 8 | 662 | 3498846 | ||
| L16926.1 | - | 8 | 1140 | |||
| U15562.1 | - | 9 | 924 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cdc25c| Gene Symbol: Cdc25c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK141737.1 | - | 9 | 958 | ||
| AK181318.1 | - | 6 | 352 | |||
| BC004669.1 | - | 8 | 662 | 3498846 | ||
| L16926.1 | - | 8 | 1140 | |||
| U15562.1 | - | 9 | 924 | |||