siRNA Details
| Gene Symbol: H1F0 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005318.3 | NP_005309.1 | 1 | 794 | ||
| GenBank mRNA | AK091372.1 | - | 2 | 584 | ||
| AK312583.1 | - | 1 | 660 | |||
| BC000145.1 | - | 1 | 641 | 3049108 | ||
| BC029046.1 | - | 1 | 641 | 5189077 | ||
| CR456502.1 | - | 1 | 472 | |||
| CR542220.1 | - | 1 | 356 | |||

H1F0
Genomic Map