siRNA Details
| Gene Symbol: Cdk6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009873.2 | NP_034003.1 | 2 | 338 | ||
| GenBank mRNA | AF132482.1 | - | 2 | 169 | ||
| AF132483.1 | - | 2 | 338 | |||
| AK078973.1 | - | 2 | 449 | |||
| AK154197.1 | - | 3 | 611 | |||
| BC119360.1 | - | 2 | 327 | 8734109 | ||
| BC120595.1 | - | 2 | 327 | 40129518 | ||

Cdk6
Genomic Map