siRNA Details
| Gene Symbol: Cdk7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009874.3 | NP_034004.2 | 2 | 216 | ||
| GenBank mRNA | AK039084.1 | - | 2 | 204 | ||
| AK145759.1 | - | 2 | 142 | |||
| AK168286.1 | - | 2 | 215 | |||
| BC004605.1 | - | 2 | 174 | 3585145 | ||
| BC068160.1 | - | 2 | 201 | 30358255 | ||
| BC141043.1 | - | 2 | 173 | 9055655 | ||
| U11822.1 | - | 2 | 179 | |||
| X74145.1 | - | 2 | 198 | |||

Cdk7
Genomic Map