siRNA Details
| Gene Symbol: Cer1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009887.2 | NP_034017.1 | 1 | 471 | ||
| GenBank mRNA | AF031896.1 | - | 1 | 485 | ||
| AF035579.1 | - | 1 | 444 | |||
| BC132431.1 | - | 1 | 454 | 40130708 | ||
| BC145993.1 | - | 1 | 452 | 40131296 | ||

Cer1
Genomic Map