siRNA Details
Gene Symbol: Chil3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | D87757.2 | - | 11 | 1262 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Chil3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | D87757.2 | - | 11 | 1262 |